EMAIL THIS PAGE TO A FRIEND
Clinica chimica acta; international journal of clinical chemistry
'True' ascorbic acid in plasma and serum: investigation of a new method.
PMID 6627672
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93285
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)
C18H12N6