EMAIL THIS PAGE TO A FRIEND
Research communications in chemical pathology and pharmacology
Prostaglandin B1 can modify the pressor responses to sympathetic nerve stimulation.
PMID 6635328
Abstract
PGB1, a metabolite of PGA1, has the ability to enhance peripheral vascular resistance and elevate blood pressure in animals whose vascular tone is low. The effect is not central in origin and apparently is not the result of changes in cholinergic or alpha-adrenoceptor sensitivity or changes in vascular smooth muscle susceptibility per se.