EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur die gesamte Hygiene und ihre Grenzgebiete
[Industrial toxicologic evaluation of vulcanizing vapors with special reference to nitrogen-containing aging-preventive agents].
PMID 6636892
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178055
N-Phenyl-2-naphthylamine, 97%
C16H13N