EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Determination of maximum permissible exposure level for the herbicide vernam in the air of a working zone].
PMID 6642234
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45704
Vernolat, PESTANAL®, analytical standard
C10H21NOS