EMAIL THIS PAGE TO A FRIEND
Health physics
A new X-ray TLD phosphor.
PMID 6643062
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
523437
Barium sulfide, 99.9%
BaS