EMAIL THIS PAGE TO A FRIEND
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie
A simple spectrophotometric assay of carboxypeptidase N (kininase I) in human serum.
PMID 6644248
Abstract
Kininase I (carboxypeptidase N; EC 3.4.17.3) consists of carboxypeptidase N1 (CN1) and carboxypeptidase N2 (CN2); these two enzymes can be differentiated by their activities towards hippuryl-L-arginine and hippuryl-L-lysine, respectively. A spectrophotometric assay for both carboxypeptidases in human serum is described and the biochemical behaviour of these enzymes investigated. The pH optima are found to be 8.4 for CN1 and CN2. The Michaelis-Menten constants are: CN1 4.59 +/- 0.03 mmol/l; CN2 37.26 +/- 3.49 mmol/l. CN2 can be inhibited by EDTA (76%), dimercaprolum (97%) and phenanthroline (98%). Diisopropylfluorophosphate has no influence on both enzymes. Elevated haemoglobin only interferes with CN1 measurements, and high bilirubin concentrations slightly alter the activity of both enzymes. High CN1 activities were found in sera of patients with sarcoidosis, and elevated CN2 activities were found in lung cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2508
Hippuryl-Arg, carboxypeptidase substrate
C15H21N5O4