EMAIL THIS PAGE TO A FRIEND
JAMA
Leads from the MMWR. Lead poisoning from Mexican folk remedies--California.
PMID 6644995
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241547
Lead oxide, red powder, 1-2 μm, 99%
O4Pb3
11536
Lead(II,IV) oxide, ≥96%
O4Pb3