EMAIL THIS PAGE TO A FRIEND
Farmakologiia i toksikologiia
[Interaction of pipolfen and suprastin with human serum albumin depending on the pH of the medium and its calcium ion content].
PMID 6653761
Abstract
Experiments made with the use of equilibrium dialysis and differential spectrophotometry have shown that pipolfen and difenhydramine interact actively with human serum albumin at pH 6.8-7.4. Calcium ions have been demonstrated to activate the interaction of pipolphen and diphenhydramine with albumin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C1915
Chloropyramine hydrochloride, analytical standard, for drug analysis
C16H20ClN3 · HCl