EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
Bio-degradation of acetates of geraniol, nerol & citronellol by P. incognita: isolation & identification of metabolites.
PMID 6671672