EMAIL THIS PAGE TO A FRIEND
Rinsho byori. The Japanese journal of clinical pathology
[Measurement of magnesium in the human sera with xylidyl-blue I. I. The effect of phosphate ion].
PMID 6672298
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248266
Xylidyl blue I
C25H20N3NaO6S