EMAIL THIS PAGE TO A FRIEND
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan
[Gas chromatographic separation of the diastereomers of labetalol as n-butylboronate].
PMID 6674493
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
19667
Butylboronic acid, for GC derivatization, ≥96.0% (T)
C4H11BO2
163244
Butylboronic acid, 97%
C4H11BO2