EMAIL THIS PAGE TO A FRIEND
Archivio Monaldi per la tisiologia e le malattie dell'apparato respiratorio
[Effect of cloperastine on muco-ciliary clearance].
PMID 6680009
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2040
Cloperastine hydrochloride, analytical standard
C20H24ClNO · HCl