EMAIL THIS PAGE TO A FRIEND
Journal of forensic sciences
The clandestine drug laboratory situation in the United States.
PMID 6680736
Abstract
Clandestine, or illegal, laboratories are operated by the criminal element to circumvent legal requirements with the goal of supplying drugs of abuse to the illicit market. Investigation of clandestine drug manufacturing laboratories is a high priority of the U.S. Drug Enforcement Administration (DEA) because elimination of these laboratories will prevent drugs of abuse from reaching the illicit drug traffic. One of the important responsibilities of forensic chemists assisting in investigations of clandestine drug laboratories is to be familiar with the methods of synthesis being used by clandestine laboratory operators. A review of clandestine laboratory seizures during the period of 1978 through 1981 will be provided to familiarize forensic chemists with current information on the types of laboratories being seized in the United States and the methods of synthesis being used.