EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Method of determining residues of a synthetic insecticide attractant in environmental objects].
PMID 6681787
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45895
Farnesyl acetate, technical, mixture of isomers
C17H28O2