EMAIL THIS PAGE TO A FRIEND
Journal of biochemical and biophysical methods
Estimation of cis-diamminedichloroplatinum (II) binding to purine bases of calf thymus DNA using [14C]methylbromphenvinphos.
PMID 6682128