EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Effect of dosanex on the immunologic reactions in animals].
PMID 6682396
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36164
Metoxuron, PESTANAL®, analytical standard
C10H13ClN2O2