EMAIL THIS PAGE TO A FRIEND
European journal of biochemistry
Purification and characterization of bovine coagulation factor XII (Hageman factor).
PMID 668690
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12072
Benzamidine, ≥95.0%
C7H8N2