EMAIL THIS PAGE TO A FRIEND
Farmakologiia i toksikologiia
[Mechanism of the microcirculatory effects of ethyl-3,5,6-tri-O-benzyl-D-glucofuranoside].
PMID 6687871
Abstract
According to the rheo- and photoplethysmography data, ethyl-3,5,6-tri-O-benzyl-D-glucofuranoside (glivenol, tribenol; 25,250 and 4000 mg/kg i.p.) markedly increases the blood content in hind limbs of rabbits, displaying a tendency towards a decrease in systemic arterial pressure and negligibly raises the body temperature when given in maximal doses, without affecting cardiac activity and respiration. The mechanism of the drug hemodynamic effect revealed is determined by activation of the blood flow in precapillary vessels. This plays an essential role in pathogenetic therapy of microcirculatory and trophic disorders in chronic venous failure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000165 Tribenoside, European Pharmacopoeia (EP) Reference Standard
C29H34O6