EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Rheumatologie
[Test of potential antiarthritic drugs on spontaneous arthritis of mice].
PMID 6689102
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0000165 Tribenoside, European Pharmacopoeia (EP) Reference Standard
C29H34O6