EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Immunocytochemistry for LHRH neurons in the arcuate nucleus area of the rat: fact or artifact?
PMID 6690601