EMAIL THIS PAGE TO A FRIEND
Toxicology letters
Metabolism of vinylcyclooctane and partition ratio between epoxide formation and cytochrome P-450 destruction.
PMID 6701911
Abstract
Vinylcyclooctane (VCO), which binds to the active site of cytochrome P-450 (P-450) giving a type I difference spectrum, has been found to form the corresponding epoxide as the main metabolite on treatment with liver microsomal monooxygenase obtained from phenobarbital-treated or untreated mice. During this metabolic process about 40% of the microsomal P-450 isozymes are destroyed, but the remainder still demethylates aminopyrine. Approx. 180 molecules of VCO are turned over and 132 of epoxyethylcyclooctane (EECO) are formed for each destructive event.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94959
Vinylcyclooctane, ≥98.0%
C10H18