EMAIL THIS PAGE TO A FRIEND
Cancer research
Inhibition by magnesium and calcium acetates of lead subacetate- and nickel acetate-induced lung tumors in strain A mice.
PMID 6704965
Abstract
The ability of the physiologically essential divalent metals calcium and magnesium to inhibit the tumorigenic activities of lead and nickel towards the lungs of strain A mice was investigated. The tumorigenic salts lead(II) subacetate and nickel(II) acetate were injected i.p. at their maximal tolerated doses (0.04 mmol/kg/injection of each metal) for a total of 24 injections, whenever possible. Calcium(II) acetate and magnesium(II) acetate were administered in the same preparation along with the lead and nickel salts at molar doses of approximately 1, 3, 10, and 30 times the maximal tolerated dose of the tumorigen. The animals were sacrificed 30 weeks after the first injection, and the lung tumors were counted. The lead and nickel salts, administered alone, each produced a significant increase in the observed number of lung adenomas per mouse. When administered with any of the doses of calcium acetate or magnesium acetate tested, neither lead subacetate nor nickel acetate showed any significant tumorigenic activity. Calcium acetate alone (total dose, 11 mmol/kg of body weight) appeared to yield a significant rise in lung adenomas observed. The results indicate an antagonism between magnesium and calcium and the tumorigenic metals nickel and lead.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
237159
Lead subacetate, ACS reagent
C4H10O8Pb3