EMAIL THIS PAGE TO A FRIEND
Dermatosen in Beruf und Umwelt. Occupation and environment
[Virostatic effect in the treatment of pityriasis rosea irritata with moroxydine-HCI].
PMID 6705693
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
278610
Moroxydine hydrochloride, 99%
C6H13N5O · HCl