EMAIL THIS PAGE TO A FRIEND
European journal of drug metabolism and pharmacokinetics
Percutaneous absorption of 5-iodo-2' deoxycytidine in the hairless rat and in man.
PMID 6714272
Abstract
A study of percutaneous absorption of the anti-herpetic agent 5-iodo-2' deoxycytidine (IDC) was performed in vitro on the rat and on man and in vivo on the rat, using samples of normal skin and skin stripped of its stratum corneum. With normal skin, absorption was 1.84 cm.h1 10(-4) for the rat and 0.33 cm.h1 10(-4) for human skin; when the stratum corneum was absent, absorption was approximately 15 times higher. The results are discussed in terms of efficacy and toxicity of the drug.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I7000
5-Iodo-2′-deoxycytidine
C9H12IN3O4