EMAIL THIS PAGE TO A FRIEND
Il Farmaco; edizione pratica
[Physical properties and availability of chloramphenicol palmitate in powdered mixtures containing microcrystalline cellulose].
PMID 6714394
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46109
Chloramphenicol palmitate, VETRANAL, analytical standard
C27H42Cl2N2O6