EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Hygienic establishment of maximum permissible concentrations of nitrilotrimethylphosphonic acid and a copper-containing complex in reservoir water].
PMID 6714698
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72568
Nitrilotri(methylphosphonic acid), ≥97.0% (T)
C3H12NO9P3