EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Conformational changes in mitochondrial aspartate aminotransferase detected by a covalently attached fluorescent probe.
PMID 6718394
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71028
Monobromo(trimethylammonio)bimane bromide, suitable for fluorescence, ≥90.0% (HPLC)
C13H19Br2N3O2