EMAIL THIS PAGE TO A FRIEND
Modern veterinary practice
Onion toxicity.
PMID 6727836
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
149225
Dipropyl disulfide, 98%
C6H14S2
W322806
Propyl disulfide, ≥97%, FG
C6H14S2