EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
The role of pyridoxal 5'-phosphate and orthophosphate in general acid-base catalysis by alpha-glucan phosphorylases.
PMID 6728843
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
464058
D-Glucal, 96%
C6H10O4