EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Lack of stringent stereospecificity in the inactivation of pyridoxal phosphate-dependent enzymes by suicide-substrates.
PMID 6728852