EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Toxicity of trichlorotoluene isomers: a 28-day feeding study in the rat.
PMID 6736562
Abstract
Groups of 10 male and 10 female rats were fed alpha,alpha,alpha-, alpha,2,6- or 2,3,6- trichlorotoluene (TCT) in their diet at 0, 0.5, 5.0, 50 or 500 ppm for 28 days. Growth rate and food consumption were not affected by treatment. No deaths occurred. Significant increases in liver weights were observed in male rats fed 5.0 and 500 ppm 2,3,6-TCT. Mild serum biochemical changes occurred in male rats. These included increased SDH activities in the groups fed 5.0 and 50 ppm alpha, alpha, alpha-TCT, and 5.0 ppm 2,3,6-TCT. Alpha, alpha, alpha-TCT at 500 ppm caused elevated LDH activities in male rats. Hepatic microsomal aminopyrine N-demethylase activities were increased in male rats fed 500 ppm alpha,2,6-TCT. Hematological parameters were not affected by treatment. Mild histological changes were seen in the liver, kidney and thyroid of treated rats. Data presented here suggest that alpha, alpha, alpha-, alpha,2,6- and 2,3,6-TCT possess a low order of oral toxicity in the rat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T56308
2,6-Dichlorobenzyl chloride, 97%
C7H5Cl3
147893
α,α,α-Trichlorotoluene, 98%
C7H5Cl3