EMAIL THIS PAGE TO A FRIEND
Teratology
An embryotoxicity study of the fungicide tridemorph and its commercial formulation Calixin.
PMID 6740510
Abstract
Tridemorph (N-tridecyl-2,6-dimethylmorpholine), the active ingredient of the commercially formulated fungicide Calixin, is a teratogen in rats and mice. The no-effect level for embryotoxic effects was 27.5 mg/kg for mice and 20.6 mg/kg for rats. By contrast, when Calixin, which contains 83% tridemorph, was administered orally at dose levels of 0.156, 0.722, and 3.909 mg/kg, no embryotoxic effects were observed in two strains of rats. Our extensive investigations, carried out under exposure conditions resembling as closely as possible those reported in another study, did not reproduce the previous findings of teratogenicity of Calixin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36199
Tridemorph, PESTANAL®, analytical standard
C19H39NO