EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
Rapid measurement of serum magnesium with a kit.
PMID 6744615
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C204
Calmagite, indicator grade
C17H14N2O5S