EMAIL THIS PAGE TO A FRIEND
European journal of immunology
The roles of host and donor cells in the rejection of skin allografts by T cell-deprived rats injected with syngeneic T cells.
PMID 6749527
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
OX-19, Reactivity: Anti-CD5, rat, 84112012