EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
19F nuclear magnetic resonance observations of aldehyde dismutation catalyzed by horse liver alcohol dehydrogenase.
PMID 6751236
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
224944
4-(Trifluoromethyl)benzaldehyde, 98%
C8H5F3O