EMAIL THIS PAGE TO A FRIEND
Japanese journal of pharmacology
Potentiation of bradykinin-induced decrease of blood pressure in dogs by SO 14,551, the disulfide metabolite of captopril.
PMID 6757503
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1091221 Captopril disulfide, United States Pharmacopeia (USP) Reference Standard
C18H28N2O6S2