EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Tyrosyl residues of two molecular forms of human urinary urokinase: their ionization mode characterized by spectrophotometric and CD titrations and reactivity with cyanuric fluoride in relation to enzymic activity.
PMID 6758771
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
28625
Cyanuric fluoride
C3F3N3