EMAIL THIS PAGE TO A FRIEND
Iyo denshi to seitai kogaku. Japanese journal of medical electronics and biological engineering
[Cancer and radiotherapy. (4). Hypoxic cell sensitizers and hyperthermia combined with radiation in cancer treatment].
PMID 6759728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N9608
4′-Nitroacetophenone, 98%
C8H7NO3