EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Sensitive fluorogenic substrates for the detection of trypsinlike proteases and pancreatic elastase.
PMID 6760743
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
275581
6-Aminoquinoline, 98%
C9H8N2