EMAIL THIS PAGE TO A FRIEND
Toxicology and applied pharmacology
Comparative gastric ulcerogenic effects of meseclazone, 5-chlorosalicylic acid and other nonsteroidal anti-inflammatory drugs following acute and repeated oral administration to rats.
PMID 6768168
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C70908
5-Chlorosalicylic acid, 98%
C7H5ClO3