EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Pre-prolactin accumulates in rat pituitary cells incubated with a threonine analog.
PMID 6771282
Abstract
The proteolytic processing of rat pre-prolactin (Pre-Prl) to prolactin in isolated hemipituitaries was inhibited by the threonine analog, beta-hydroxynorvaline (Hnv). Pre-Prl accumulated as a major labeled intracellular product, even during pulse labeling longer than 2 h, but it was not secreted. Inhibition of Pre-Prl processing may result from incorporation of Hnv at the Pre-Prl cleavage site. This system offers the opportunity to study the disposition of an accumulated pre-protein in cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4002
DL-3-Hydroxynorvaline, ≥98% (TLC)
C5H11NO3