EMAIL THIS PAGE TO A FRIEND
The American journal of physiology
Role of urinary bladder in osmoregulation in marine teleost, Opsanus tau.
PMID 677339
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248614
Ammonium persulfate, ACS reagent, ≥98.0%
H8N2O8S2