EMAIL THIS PAGE TO A FRIEND
Clinical chemistry
[Bis(2-hydroxyethyl)amino]tris(hydroxymethyl)methane is an effective buffer for creatine kinase assays.
PMID 6773703
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14879
BIS-TRIS, BioUltra, ≥99.0% (NT)
C8H19NO5