EMAIL THIS PAGE TO A FRIEND
Acta histochemica
[1-methoxy-5-methylphenazinium-methylsulfate--a light stabile redox mediator in the histochemistry (author's transl)].
PMID 6774587
Abstract
1-Methoxy-5-methylphenazinium methyl sulfate (MPMS) was tested for use as redox mediator in histochemistry of dehydrogenases. Aqueous solutions of MPMS are absolute lightstable. Like PMS or Meldola Blue the succinate dehydrogenase reaction was increased by MPMS. In an non-enzymatic NADH-NBT-test the reaction rate of MPMS exceeds the rates of PMS or Meldola Blue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M8640
1-Methoxy-5-methylphenazinium methyl sulfate, ≥95%
C15H16N2O5S