EMAIL THIS PAGE TO A FRIEND
Biochemistry
Comparison of the beta-galactosidase conformations induced by D-galactal and by magnesium ions.
PMID 6774745
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
462233
D-Galactal, 95%
C6H10O4