EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Characterization of N-methylphenylethylamine and N-methylphenylethanolamine as substrates for type A and type B monoamine oxidase.
PMID 6775642
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
209848
α-(Methylaminomethyl)benzyl alcohol, 99%
C9H13NO
M68423
N-Methyl-phenethylamine, 99%
C9H13N