EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Comparative study on accumulation and elimination of hexachlorobiphenyls and decachlorobiphenyl in mice.
PMID 6775714