EMAIL THIS PAGE TO A FRIEND
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes
Degradation of carboxin (Vitavax) and oxycarboxin (Plantvax) by Pseudomonas aeruginosa isolated from soil.
PMID 6776177
Abstract
Pseudomonas aeruginosa capable of utilizing carboxin and oxycarboxin as sole sources of carbon and nitrogen was isolated from red sandy loam soil perfused with the solutions of these fungicides. The bacterium hydrolyzed oxycarboxin via the intermediate compound 2-(vinylsulphonyl) acetanilide liberating 2- (2-hydroxyethylsulphonyl) acetic acid and aminophenol, whereas carboxin was first oxidized to its sulphoxide and then to its sulphone before hydrolysis. Further hydrolysis of aminophenol by the organism resulted in the accumulation of ammonium which was partly oxidized to nitrite. Nitrite accumulated in the medium without further oxidation to nitrate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36185
Oxycarboxine, PESTANAL®, analytical standard
C12H13NO4S