EMAIL THIS PAGE TO A FRIEND
Journal of steroid biochemistry
Steroid sulphate secretion by the human ovary in vitro: influence of gonadotrophins, cyclofenil and clomiphene.
PMID 6780729
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E9505
β-Estradiol 3-sulfate sodium salt, ≥93%
C18H23NaO5S