EMAIL THIS PAGE TO A FRIEND
MMWR. Morbidity and mortality weekly report
Occupationally related neurologic abnormalities--Massachusetts.
PMID 6790924
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
234486
3-(Dimethylamino)propionitrile, 98%
C5H10N2